Assay Details
| Gene Symbol: LOC101121661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012129529.3 | XP_011984919.2 | 5 - 6 | 832 | 98 | |
| XM_012129535.2 | XP_011984925.2 | 5 - 6 | 627 | 98 | ||
| XM_027964634.1 | XP_027820435.1 | 5 - 6 | 836 | 98 | ||
| Gene Symbol: LOC114113035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964632.1 | XP_027820433.1 | 5 - 6 | 95 | ||
More Information
Set Membership: |
|

LOC101121661
Genomic Map