Assay Details
| Gene Symbol: STRADA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161884.1 | NP_001155356.1 | 6 - 7 | 681 | 62 | |
| XM_012185072.3 | XP_012040462.1 | 10 - 11 | 1007 | 62 | ||
| XM_012185076.3 | XP_012040466.1 | 10 - 11 | 1025 | 62 | ||
| XM_012185079.3 | XP_012040469.1 | 10 - 11 | 983 | 62 | ||
| XM_012185082.3 | XP_012040472.1 | 10 - 11 | 1001 | 62 | ||
| XM_012185083.3 | XP_012040473.1 | 8 - 9 | 918 | 62 | ||
| XM_012185089.3 | XP_012040479.1 | 8 - 9 | 894 | 62 | ||
| XM_012185092.3 | XP_012040482.1 | 8 - 9 | 912 | 62 | ||
| XM_012185094.3 | XP_012040484.1 | 8 - 9 | 805 | 62 | ||
| XM_012185096.3 | XP_012040486.1 | 7 - 8 | 837 | 62 | ||
| XM_027973934.1 | XP_027829735.1 | 10 - 11 | 1171 | 62 | ||
| XM_027973935.1 | XP_027829736.1 | 10 - 11 | 1008 | 62 | ||
| XM_027973936.1 | XP_027829737.1 | 10 - 11 | 1010 | 62 | ||
| XM_027973937.1 | XP_027829738.1 | 7 - 8 | 3080 | 62 | ||
| GenBank mRNA | FJ943998.1 | - | 6 - 7 | 681 | 62 | |
More Information
Set Membership: |
|

STRADA
Genomic Map