Assay Details
| Gene Symbol: CDIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010430.4 | XP_004010479.2 | 6 - 7 | 632 | 71 | |
| XM_012181160.3 | XP_012036550.2 | 6 - 7 | 632 | 71 | ||
| XM_012181161.3 | XP_012036551.2 | 6 - 7 | 632 | 71 | ||
| XM_012181162.3 | XP_012036552.2 | 6 - 7 | 632 | 71 | ||
| XM_012181163.3 | XP_012036553.2 | 4 - 5 | 521 | 71 | ||
| XM_027971655.1 | XP_027827456.1 | 6 - 7 | 632 | 71 | ||
| XM_027971656.1 | XP_027827457.1 | 6 - 7 | 346 | 71 | ||

CDIN1
Genomic Map