Assay Details
| Gene Symbol: AP5M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971889.1 | XP_027827690.1 | 2 - 3 | 996 | 77 | |
| XM_027971890.1 | XP_027827691.1 | 2 - 3 | 996 | 77 | ||
| XM_027971891.1 | XP_027827692.1 | 2 - 3 | 1001 | 77 | ||
More Information
Set Membership: |
|

AP5M1
Genomic Map