Assay Details
| Gene Symbol: PRKAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097822.3 | XP_011953212.1 | 2 - 3 | 376 | 62 | |
| XM_012097823.3 | XP_011953213.1 | 2 - 3 | 292 | 62 | ||
| XM_027956524.1 | XP_027812325.1 | 1 - 2 | 540 | 62 | ||
More Information
Set Membership: |
|

PRKAB1
Genomic Map