Assay Details
| Gene Symbol: TMEM9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101041.2 | XP_014956527.2 | 4 - 5 | 547 | 66 | |
| XM_015101042.2 | XP_014956528.1 | 3 - 4 | 440 | 66 | ||
More Information
Set Membership: |
|

TMEM9B
Genomic Map