Assay Details
| Gene Symbol: SIGMAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012154855.3 | XP_012010245.3 | 2 - 3 | 471 | 91 | |
| XM_015093060.2 | XP_014948546.2 | 2 - 3 | 353 | 91 | ||
More Information
Set Membership: |
|

SIGMAR1
Genomic Map