Assay Details
| Gene Symbol: MIF4GD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974065.1 | XP_027829866.1 | 3 - 4 | 337 | 61 | |
| XM_027974066.1 | XP_027829867.1 | 3 - 4 | 323 | 61 | ||
More Information
Set Membership: |
|

MIF4GD
Genomic Map