Assay Details
| Gene Symbol: SORD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010651.4 | XP_004010700.1 | 4 - 5 | 579 | 73 | |
| XM_027971863.1 | XP_027827664.1 | 4 - 5 | 569 | 73 | ||
More Information
Set Membership: |
|

SORD
Genomic Map