Assay Details
| Gene Symbol: AIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018957.4 | XP_004019006.1 | 2 - 3 | 202 | 70 | |
| XM_027958854.1 | XP_027814655.1 | 2 - 3 | 203 | 70 | ||
More Information
Set Membership: |
|

AIF1
Genomic Map