Assay Details
| Gene Symbol: GGTA2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009764.1 | NP_001009764.1 | 1 - 2 | 84 | 101 | |
| XM_012159476.3 | XP_012014866.1 | 3 - 4 | 280 | 101 | ||
| XM_027965748.1 | XP_027821549.1 | 3 - 4 | 709 | 101 | ||
| XM_027965749.1 | XP_027821550.1 | 3 - 4 | 486 | 101 | ||
| XM_027965750.1 | XP_027821551.1 | 4 - 5 | 581 | 101 | ||
| GenBank mRNA | AY447039.1 | - | 1 - 2 | 84 | 101 | |
More Information
Set Membership: |
|

GGTA2P
Genomic Map