Assay Details
| Gene Symbol: TPM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004275.4 | XP_004004324.1 | 7 - 8 | 1740 | 63 | |
| XM_004004276.3 | XP_004004325.1 | 7 - 8 | 834 | 63 | ||
| XM_004004277.3 | XP_004004326.1 | 7 - 8 | 834 | 63 | ||
| XM_004004278.4 | XP_004004327.1 | 7 - 8 | 815 | 63 | ||
More Information
Set Membership: |
|

TPM2
Genomic Map