Assay Details
| Gene Symbol: TPPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970078.1 | XP_027825879.1 | 3 - 4 | 1994 | 54 | |
| XM_027970079.1 | XP_027825880.1 | 4 - 5 | 1851 | 54 | ||
| XM_027970080.1 | XP_027825881.1 | 4 - 5 | 1458 | 54 | ||
| XM_027970082.1 | XP_027825883.1 | 4 - 5 | 493 | 54 | ||
| XM_027970083.1 | XP_027825884.1 | 4 - 5 | 468 | 54 | ||
| XM_027970084.1 | XP_027825885.1 | 3 - 4 | 404 | 54 | ||
More Information
Set Membership: |
|

TPPP2
Genomic Map