Assay Details
| Gene Symbol: DPEP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977763.1 | XP_027833564.1 | 6 - 7 | 827 | 54 | |
| XM_027977764.1 | XP_027833565.1 | 6 - 7 | 806 | 54 | ||
| XM_027977765.1 | XP_027833566.1 | 6 - 7 | 832 | 54 | ||
More Information
Set Membership: |
|

DPEP1
Genomic Map