Assay Details
| Gene Symbol: ALDOB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009809.1 | NP_001009809.1 | 4 - 5 | 448 | 65 | |
| XM_012111144.2 | XP_011966534.1 | 5 - 6 | 778 | 65 | ||
| GenBank mRNA | Z29372.1 | - | 448 | 65 | ||
More Information
Set Membership: |
|

ALDOB
Genomic Map