Assay Details
| Gene Symbol: PDZK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002409.4 | XP_004002458.2 | 6 - 7 | 1084 | 79 | |
| XM_012179902.3 | XP_012035292.2 | 6 - 7 | 1124 | 79 | ||
| XM_012179905.3 | XP_012035295.2 | 6 - 7 | 1226 | 79 | ||
| XM_027973920.1 | XP_027829721.1 | 6 - 7 | 1074 | 79 | ||
More Information
Set Membership: |
|

PDZK1
Genomic Map