Assay Details
| Gene Symbol: ANKRD46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011810.3 | XP_004011859.2 | 3 - 4 | 548 | 102 | |
| XM_012184053.2 | XP_012039443.2 | 2 - 3 | 475 | 102 | ||
| XM_027973174.1 | XP_027828975.1 | 5 - 6 | 1154 | 102 | ||
| XM_027973175.1 | XP_027828976.1 | 5 - 6 | 1154 | 102 | ||
More Information
Set Membership: |
|

ANKRD46
Genomic Map