Assay Details
| Gene Symbol: SERPINC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009393.1 | NP_001009393.1 | 4 - 5 | 812 | 66 | |
| GenBank mRNA | X68287.1 | - | 812 | 66 | ||
More Information
Set Membership: |
|

SERPINC1
Genomic Map