Assay Details
| Gene Symbol: SRSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195316.1 | NP_001182245.1 | 2 - 3 | 343 | 81 | |
| XM_012100410.2 | XP_011955800.1 | 3 - 4 | 476 | 81 | ||
| GenBank mRNA | EU370536.1 | - | 2 - 3 | 343 | 81 | |
More Information
Set Membership: |
|

SRSF3
Genomic Map