Assay Details
| Gene Symbol: LAMTOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002602.4 | XP_004002651.1 | 3 - 4 | 426 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LAMTOR2| Gene Symbol: LAMTOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002602.4 | XP_004002651.1 | 3 - 4 | 426 | 60 | |
Set Membership: |
|