Assay Details
| Gene Symbol: MLX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098794.2 | XP_014954280.2 | 6 - 7 | 668 | 62 | |
| XM_027974496.1 | XP_027830297.1 | 4 - 5 | 658 | 62 | ||
| XM_027974497.1 | XP_027830298.1 | 5 - 6 | 569 | 62 | ||
| XM_027974498.1 | XP_027830299.1 | 6 - 7 | 543 | 62 | ||
| XM_027974499.1 | XP_027830300.1 | 5 - 6 | 447 | 62 | ||
More Information
Set Membership: |
|

MLX
Genomic Map