Assay Details
| Gene Symbol: MRPL45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012821.3 | XP_004012870.1 | 6 - 7 | 760 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRPL45| Gene Symbol: MRPL45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012821.3 | XP_004012870.1 | 6 - 7 | 760 | 55 | |
Set Membership: |
|