Assay Details
| Gene Symbol: PPP1CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012102181.3 | XP_011957571.1 | 3 - 4 | 531 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1CA| Gene Symbol: PPP1CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012102181.3 | XP_011957571.1 | 3 - 4 | 531 | 64 | |
Set Membership: |
|