Assay Details
| Gene Symbol: OXA1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010354.4 | XP_004010403.1 | 7 - 8 | 953 | 64 | |
| XM_012181018.3 | XP_012036408.1 | 7 - 8 | 944 | 64 | ||
| XM_012181019.2 | XP_012036409.1 | 6 - 7 | 750 | 64 | ||
More Information
Set Membership: |
|

OXA1L
Genomic Map