Assay Details
| Gene Symbol: ADPRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015091860.2 | XP_014947346.2 | 4 - 5 | 716 | 102 | |
| XM_027967823.1 | XP_027823624.1 | 4 - 5 | 2155 | 102 | ||
More Information
Set Membership: |
|

ADPRS
Genomic Map