Assay Details
| Gene Symbol: ACAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960802.1 | XP_027816603.1 | 8 - 9 | 1189 | 71 | |
| XM_027960803.1 | XP_027816604.1 | 8 - 9 | 1225 | 71 | ||
More Information
Set Membership: |
|

ACAA2
Genomic Map