Assay Details
| Gene Symbol: BAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019650.4 | XP_004019699.1 | 3 - 4 | 494 | 62 | |
| XM_012102095.3 | XP_011957485.1 | 3 - 4 | 564 | 62 | ||
| XM_012102096.3 | XP_011957486.1 | 3 - 4 | 527 | 62 | ||
| GenBank mRNA | KY320499.1 | - | 3 - 4 | 458 | 62 | |
More Information
Set Membership: |
|

BAD
Genomic Map