Assay Details
| Gene Symbol: LOC114112693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963843.1 | XP_027819644.1 | 1 - 2 | 92 | 105 | |
| Gene Symbol: SSBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970492.1 | XP_027826293.1 | 6 - 7 | 625 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027963843.1 transcripts.
Set Membership: |
|

LOC114112693
Genomic Map