Assay Details
| Gene Symbol: SLC16A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002335.4 | XP_004002384.1 | 3 - 4 | 565 | 89 | |
| GenBank mRNA | AJ315929.1 | - | 2 - 3 | 280 | 89 | |
More Information
Set Membership: |
|

SLC16A1
Genomic Map