Assay Details
| Gene Symbol: LOC101107455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975952.1 | XP_027831753.1 | 1 - 2 | 1145 | 86 | |
| Gene Symbol: LOC114117277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976092.1 | XP_027831893.1 | 2 - 3 | 275 | 86 | |

LOC101107455
Genomic Map