Assay Details
| Gene Symbol: PAEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009366.1 | NP_001009366.1 | 3 - 4 | 354 | 63 | |
| GenBank mRNA | X04520.1 | - | 354 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PAEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009366.1 | NP_001009366.1 | 3 - 4 | 354 | 63 | |
| GenBank mRNA | X04520.1 | - | 354 | 63 | ||
Set Membership: |
|