Assay Details
| Gene Symbol: CDK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127269.1 | NP_001120741.1 | 3 - 4 | 519 | 65 | |
| XM_012158548.3 | XP_012013938.2 | 4 - 5 | 738 | 65 | ||
| GenBank mRNA | EU525168.1 | - | 3 - 4 | 405 | 65 | |
| EU626223.1 | - | 3 - 4 | 519 | 65 | ||
| FJ943997.1 | - | 3 - 4 | 519 | 65 | ||
More Information
Set Membership: |
|

CDK4
Genomic Map