Assay Details
| Gene Symbol: UBE2H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008057.4 | XP_004008106.1 | 6 - 7 | 869 | 67 | |
| XM_015095381.2 | XP_014950867.1 | 6 - 7 | 554 | 67 | ||
| XM_015095383.2 | XP_014950869.1 | 5 - 6 | 313 | 67 | ||
More Information
Set Membership: |
|

UBE2H
Genomic Map