Assay Details
| Gene Symbol: B4GALT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980218.1 | XP_027836019.1 | 6 - 7 | 1093 | 61 | |
| XM_027980219.1 | XP_027836020.1 | 5 - 6 | 1022 | 61 | ||
More Information
Set Membership: |
|

B4GALT4
Genomic Map