Assay Details
| Gene Symbol: SPRYD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973568.1 | XP_027829369.1 | 2 - 3 | 450 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPRYD7| Gene Symbol: SPRYD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973568.1 | XP_027829369.1 | 2 - 3 | 450 | 86 | |
Set Membership: |
|