Assay Details
| Gene Symbol: SNUPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017838.4 | XP_004017887.1 | 8 - 9 | 873 | 72 | |
| XM_012098663.3 | XP_011954053.1 | 8 - 9 | 850 | 72 | ||
More Information
Set Membership: |
|

SNUPN
Genomic Map