Assay Details
| Gene Symbol: PSMA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139446.2 | NP_001132918.1 | 1 - 2 | 160 | 86 | |
| GenBank mRNA | FJ223570.1 | - | 1 - 2 | 161 | 86 | |
More Information
Set Membership: |
|

PSMA6
Genomic Map