Assay Details
| Gene Symbol: DDX55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017319.4 | XP_004017368.4 | 8 - 9 | 863 | 66 | |
| XM_012097695.3 | XP_011953085.3 | 8 - 9 | 863 | 66 | ||
More Information
Set Membership: |
|

DDX55
Genomic Map