Assay Details
| Gene Symbol: MINDY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971795.1 | XP_027827596.1 | 5 - 6 | 1325 | 79 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MINDY2| Gene Symbol: MINDY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971795.1 | XP_027827596.1 | 5 - 6 | 1325 | 79 | |