Assay Details
| Gene Symbol: LOC114117808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977572.1 | XP_027833373.1 | 4 - 5 | 805 | 96 | |
| XM_027977575.1 | XP_027833376.1 | 3 - 4 | 712 | 96 | ||
| Gene Symbol: UAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977559.1 | XP_027833360.1 | 4 - 5 | 855 | 96 | |
| XM_027977565.1 | XP_027833366.1 | 5 - 6 | 1060 | 96 | ||
| XM_027977570.1 | XP_027833371.1 | 4 - 5 | 855 | 96 | ||

LOC114117808
Genomic Map