Assay Details
| Gene Symbol: ZNF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020433.4 | XP_004020482.2 | 2 - 3 | 591 | 82 | |
| XM_012103348.2 | XP_011958738.2 | 2 - 3 | 720 | 82 | ||
More Information
Set Membership: |
|

ZNF24
Genomic Map