Assay Details
| Gene Symbol: PCGF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971353.1 | XP_027827154.1 | 4 - 5 | 608 | 68 | |
| XM_027971354.1 | XP_027827155.1 | 4 - 5 | 594 | 68 | ||
| XM_027971355.1 | XP_027827156.1 | 4 - 5 | 597 | 68 | ||
| XM_027971356.1 | XP_027827157.1 | 4 - 5 | 573 | 68 | ||
| XM_027971357.1 | XP_027827158.1 | 3 - 4 | 428 | 68 | ||
More Information
Set Membership: |
|

PCGF3
Genomic Map