Assay Details
| Gene Symbol: LOC101121607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010760.4 | XP_004010809.1 | 1 - 2 | 218 | 66 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101121607| Gene Symbol: LOC101121607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010760.4 | XP_004010809.1 | 1 - 2 | 218 | 66 | |