Assay Details
| Gene Symbol: LRRC46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974642.1 | XP_027830443.1 | 7 - 8 | 656 | 57 | |
| XM_027974643.1 | XP_027830444.1 | 6 - 7 | 814 | 57 | ||
| XM_027974644.1 | XP_027830445.1 | 6 - 7 | 829 | 57 | ||
| XM_027974645.1 | XP_027830446.1 | 6 - 7 | 814 | 57 | ||
More Information
Set Membership: |
|

LRRC46
Genomic Map