Assay Details
| Gene Symbol: CCKBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012095888.3 | XP_011951278.2 | 2 - 3 | 510 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCKBR| Gene Symbol: CCKBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012095888.3 | XP_011951278.2 | 2 - 3 | 510 | 66 | |
Set Membership: |
|