Assay Details
| Gene Symbol: PIPOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012513.4 | XP_004012562.1 | 2 - 3 | 333 | 68 | |
| XM_012185411.3 | XP_012040801.1 | 2 - 3 | 333 | 68 | ||
More Information
Set Membership: |
|

PIPOX
Genomic Map