Assay Details
| Gene Symbol: EGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142506.1 | NP_001135978.1 | 1 - 2 | 316 | 80 | |
| GenBank mRNA | EU552504.1 | - | 1 - 2 | 316 | 80 | |
More Information
Set Membership: |
|

EGR1
Genomic Map