Assay Details
| Gene Symbol: PLPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970123.1 | XP_027825924.1 | 4 - 5 | 608 | 75 | |
| XM_027970124.1 | XP_027825925.1 | 4 - 5 | 569 | 75 | ||
More Information
Set Membership: |
|

PLPP2
Genomic Map