Assay Details
| Gene Symbol: VPS35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014966.4 | XP_004015015.2 | 6 - 7 | 811 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VPS35| Gene Symbol: VPS35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014966.4 | XP_004015015.2 | 6 - 7 | 811 | 79 | |
Set Membership: |
|