Assay Details
| Gene Symbol: PQBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963646.1 | XP_027819447.1 | 3 - 4 | 369 | 65 | |
| XM_027963647.1 | XP_027819448.1 | 3 - 4 | 413 | 65 | ||
| XM_027963648.1 | XP_027819449.1 | 3 - 4 | 352 | 65 | ||
| XM_027963649.1 | XP_027819450.1 | 3 - 4 | 363 | 65 | ||
| XM_027963650.1 | XP_027819451.1 | 3 - 4 | 342 | 65 | ||
More Information
Set Membership: |
|

PQBP1
Genomic Map